Please login to your online account to display your discounted pricing

Cyclododecanone, 99+%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  110970050

 View more versions of this product

Catalog No. AC110970050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 182.31
Assay ≥99.0%
Physical Form Adhering Crystals
Quantity 5g
Color Colorless to white
Melting Point 59° to 61°C
TSCA TSCA
EINECS Number 212-595-6
Beilstein 07, II, 48
Name Note 99+%
Molecular Formula C12H22O
MDL Number MFCD00003722
Assay Percent Range 99+%
Chemical Name or Material Cyclododecanone
CAS 830-13-7
Linear Formula C12H122(=O)
Health Hazard 1 GHS Signal Word: Warning
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Cyclododecanone
Name Note 99+%
CAS 830-13-7
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C12H22O
Linear Formula C12H122(=O)
Formula Weight 182.31
MDL Number MFCD00003722
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic to aquatic life with long lasting effects.

SDS & Safety Documents