missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclododecanone, 99+%, ACROS Organics™

$58.85 - $58.85

Chemical Identifiers

CAS 830-13-7
Molecular Formula C12H22O
Molecular Weight (g/mol) 182.307
MDL Number MFCD00003722
InChI Key SXVPOSFURRDKBO-UHFFFAOYSA-N
Synonym cyclododecan-1-one, dsstox_cid_7322, dsstox_rid_78407, dsstox_gsid_27322, cyclododecanon, cdon, cyclododecanone, acmc-209pr1, ksc448s4j
PubChem CID 13246
IUPAC Name cyclododecanone
SMILES C1CCCCCC(=O)CCCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC110975000 SDS View Documents Acros Organics
110975000
500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC110971000 SDS View Documents Acros Organics
110971000
100g Each for $58.85
Specifications

Chemical Identifiers

830-13-7
182.307
SXVPOSFURRDKBO-UHFFFAOYSA-N
13246
C1CCCCCC(=O)CCCCC1
C12H22O
MFCD00003722
cyclododecan-1-one, dsstox_cid_7322, dsstox_rid_78407, dsstox_gsid_27322, cyclododecanon, cdon, cyclododecanone, acmc-209pr1, ksc448s4j
cyclododecanone

Specifications

118°C
Glass bottle
Colorless to White
500g
99+%
C12H22(=O)
07, II, 48
Solubility in water: insoluble.
C1CCCCCC(=O)CCCCC1
182.307
182.31
≥99%
Authentic
277.0°C
59.0°C to 61.0°C
830-13-7
C12H22O
MFCD00003722
cyclododecan-1-one, dsstox_cid_7322, dsstox_rid_78407, dsstox_gsid_27322, cyclododecanon, cdon, cyclododecanone, acmc-209pr1, ksc448s4j
SXVPOSFURRDKBO-UHFFFAOYSA-N
cyclododecanone
13246
Adhering Crystals
Cyclododecanone, 99+%
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 212-595-6

TSCA : TSCA