Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclododecanone, 99%

Chemical Identifiers

Chemical Name or Material Cyclododecanone
CAS 830-13-7
Molecular Formula C12H22O
Formula Weight 182.31
MDL Number MFCD00003722
UN Number UN3077
Beilstein 508621
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1156336

  SDS
alfa aesar™
L1156336
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1156322

  SDS
alfa aesar™
L1156322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 182.31
Chemical Name or Material Cyclododecanone
Molecular Formula C12H22O
MDL Number MFCD00003722
TSCA TSCA
Odor Camphor-like
Assay 99%
UN Number UN3077
Beilstein 508621
Melting Point 59° to 62°C
Flash Point 137°C (278°F)
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
CAS 830-13-7
Boiling Point 277°C
Quantity 500g