missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cyclododecanol, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 110965000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cyclododecanol | |
1724-39-6 | |
White | |
138°C | |
98% min. (GC) | |
C12H24O | |
MFCD00003717 | |
unii-0z645y4jts, dsstox_cid_7429, dsstox_rid_78451, dsstox_gsid_27429, acmc-1bprz | |
SFVWPXMPRCIVOK-UHFFFAOYSA-N | |
cyclododecanol | |
15595 | |
98+% |
99% | |
76.0°C to 79.0°C | |
272.7°C | |
Authentic | |
Glass bottle | |
C12H23OH | |
500 g | |
Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents,19.2g/100g benzene,58.5g/100g methanol,33.2g/100g chloroform | |
C1CCCCCC(CCCCC1)O | |
184.32 | |
184.32 | |
Crystalline Solid |
Chemical Identifiers
1724-39-6 | |
184.32 | |
SFVWPXMPRCIVOK-UHFFFAOYSA-N | |
15595 | |
C1CCCCCC(CCCCC1)O |
C12H24O | |
MFCD00003717 | |
unii-0z645y4jts, dsstox_cid_7429, dsstox_rid_78451, dsstox_gsid_27429, acmc-1bprz | |
cyclododecanol |
Safety and Handling
GHS H Statement
Toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
GHS Signal Word: Warning
EINECSNumber : 217-031-2
TSCA : TSCA
RUO – Research Use Only