Please login to your online account to display your discounted pricing

Cyclododecanol 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  110960050

 View more versions of this product

Catalog No. AC110960050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Solid
Assay ≥98.5%
Formula Weight 184.32
Quantity 5g
Color White
TSCA TSCA
EINECS Number 217-031-2
Melting Point 76° to 79°C
Infrared Spectrum Authentic
Name Note 99%
Assay Percent Range 99%
Linear Formula C12H23OH
MDL Number MFCD00003717
CAS 1724-39-6
Health Hazard 2 GHS H Statement:
Toxic to aquatic life with long lasting effects.
Chemical Name or Material Cyclododecanol
Packaging Glass bottle
Molecular Formula C12H24O
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Cyclododecanol
Name Note 99%
CAS 1724-39-6
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C12H24O
Linear Formula C12H23OH
Formula Weight 184.32
MDL Number MFCD00003717
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic to aquatic life with long lasting effects.

SDS & Safety Documents