missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclododecanol 99%, ACROS Organics™

$162.44 - $162.44

Chemical Identifiers

CAS 1724-39-6
Molecular Formula C12H24O
Molecular Weight (g/mol) 184.323
MDL Number MFCD00003717
InChI Key SFVWPXMPRCIVOK-UHFFFAOYSA-N
Synonym unii-0z645y4jts, dsstox_cid_7429, dsstox_rid_78451, dsstox_gsid_27429, acmc-1bprz
PubChem CID 15595
IUPAC Name cyclododecanol
SMILES C1CCCCCC(CCCCC1)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC110965000 SDS View Documents Acros Organics
110965000
500g Glass bottle Each for $162.44
AC110961000 SDS View Documents Acros Organics
110961000
100g Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC110960050 SDS View Documents Acros Organics
110960050
5g Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1724-39-6
184.323
SFVWPXMPRCIVOK-UHFFFAOYSA-N
15595
C1CCCCCC(CCCCC1)O
C12H24O
MFCD00003717
unii-0z645y4jts, dsstox_cid_7429, dsstox_rid_78451, dsstox_gsid_27429, acmc-1bprz
cyclododecanol

Specifications

138°C
Glass bottle
White
500g
99%
C12H23OH
unii-0z645y4jts, dsstox_cid_7429, dsstox_rid_78451, dsstox_gsid_27429, acmc-1bprz
C1CCCCCC(CCCCC1)O
184.323
184.32
99%
Authentic
272.7°C
76.0°C to 79.0°C
1724-39-6
C12H24O
MFCD00003717
SFVWPXMPRCIVOK-UHFFFAOYSA-N
cyclododecanol
15595
Crystalline Solid
Cyclododecanol, 99%
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 217-031-2

TSCA : TSCA