Please login to your online account to display your discounted pricing

α-Cyclodextrin (White Powder), Fisher BioReagents


Quantity 25g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP250125

 View more versions of this product

Catalog No. BP250125

  • / Each
  • / Case of 7 Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material α-Cyclodextrin
CAS 10016-20-3
ChemAlert Storage Symbol Gray
Recommended Storage RT
Identification Pass Test
Loss on Drying 13% max.
Physical Form Powder/Solid
Packaging Amber Glass
Molecular Formula C36H60O32
Formula Weight 972.9
Merck Index 15, 2710
Synonym α-CD
Specific Rotation 143 to 153° (+ or -)
Color White
Melting Point 278°C
Quantity 25g

Specifications

Chemical Name or Material α-Cyclodextrin
CAS 10016-20-3
Physical Form Powder/Solid
Molecular Formula C36H60O32
Formula Weight 972.9
Synonym α-CD

  • A cyclic polysaccharide comprised of 6 glucose units that improves the solubility of hydrophobic molecules
  • Improves the solubility of nonpolar macromolecules such as fatty acids, lipids, and cholesterol in aqueous media
  • Also used for selective precipitation of enantiomeric, positional or structural isomers in chromatrographic separations and purification methods

Accessories
 

RT

SDS & Safety Documents