missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

α-Cyclodextrin (White Powder), Fisher BioReagents

$306.58 - $306.58

Chemical Identifiers

CAS 10016-20-3
Molecular Formula C36H60O30
Molecular Weight (g/mol) 972.846
InChI Key HFHDHCJBZVLPGP-UHFFFAOYSA-N
Synonym .alpha.-cycloamylose, .alpha.-cyclodextrin, .alpha.-dextrin, .alpha.-schardinger dextrin, bdbm4, cyclohexaamylose, cyclomaltohexose, dextrin, .alpha.-cyclo, schardinger alpha-dextrin
PubChem CID 320760
SMILES C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(O2)C(C7O)O)CO)CO)CO)CO)CO)O)O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP25015
SDS View Documents Promotion Details
Fisher BioReagents
BP25015
5g Amber Glass
Each for $306.58
Case of 9 Each for $2,759.22
 
Description

Description

α-Cyclodextrin is used in chromatographic separations and purification methods. Useful for selective precipitation of enantiomeric, positional or structural isomers.
Specifications

Chemical Identifiers

10016-20-3
972.846
.alpha.-cycloamylose, .alpha.-cyclodextrin, .alpha.-dextrin, .alpha.-schardinger dextrin, bdbm4, cyclohexaamylose, cyclomaltohexose, dextrin, .alpha.-cyclo, schardinger alpha-dextrin
C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(O2)C(C7O)O)CO)CO)CO)CO)CO)O)O)O
C36H60O30
HFHDHCJBZVLPGP-UHFFFAOYSA-N
320760

Specifications

10016-20-3
278°C
Pass Test
Amber Glass
5g
HFHDHCJBZVLPGP-UHFFFAOYSA-N
143 to 153° (+ or -)
320760
Powder/Solid
White
13% max.
C36H60O30
15, 2710
.alpha.-cycloamylose, .alpha.-cyclodextrin, .alpha.-dextrin, .alpha.-schardinger dextrin, bdbm4, cyclohexaamylose, cyclomaltohexose, dextrin, .alpha.-cyclo, schardinger alpha-dextrin
C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(O2)C(C7O)O)CO)CO)CO)CO)CO)O)O)O
972.846
972.9
α-Cyclodextrin
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS
Product Certifications