missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

β-Cyclodextrin hydrate, 99%, ACROS Organics™

$81.64 - $225.06

Chemical Identifiers

CAS 68168-23-0
Molecular Formula C42H72O36
Molecular Weight (g/mol) 1153.002
MDL Number MFCD00150811
InChI Key JNSAKCOAFBFODP-ZQOBQRRWSA-N
Synonym beta-cyclodextrin hydrate, schardinger, a-dextrin hydrate, beta-cyclodextrin hydrate, beta-cyclodextrin hydrate, puriss hplc
PubChem CID 53486154
SMILES C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(O2)C(C8O)O)CO)CO)CO)CO)CO)CO)O)O)O.O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC227280250 SDS View Documents Acros Organics
227280250
25g Glass bottle Each for $81.64
AC227280050 SDS View Documents Acros Organics
227280050
5g Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC227281000 SDS View Documents Acros Organics
227281000
100g Glass bottle Each for $225.06
Description

Description

Chelation/Complexation reagent
Specifications

Chemical Identifiers

68168-23-0
1153.002
JNSAKCOAFBFODP-ZQOBQRRWSA-N
53486154
C42H72O36
MFCD00150811
beta-cyclodextrin hydrate, schardinger, a-dextrin hydrate, beta-cyclodextrin hydrate, beta-cyclodextrin hydrate, puriss hplc
C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(O2)C(C8O)O)CO)CO)CO)CO)CO)CO)O)O)O.O

Specifications

(1% aq. soln.) clear, colorless
Glass bottle
+155° to +167° (25°C, 589nm) (c=1, H20) on dry substance
14% Max. (K.F.)
25g
68168-23-0
100.0
C42H72O36
06,151; 08,133; 09,129; 12,151; 15,107
beta-cyclodextrin hydrate, schardinger, a-dextrin hydrate, beta-cyclodextrin hydrate, beta-cyclodextrin hydrate, puriss hplc
C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(O2)C(C8O)O)CO)CO)CO)CO)CO)CO)O)O)O.O
53486154
Crystalline Powder
β-Cyclodextrin hydrate, 99%
Authentic
5ppm toluene max.
0.1 % max.
White
5.0 to 8.0 (1% aq. soln.)
98.5
99%
MFCD00150811
14, 2718
JNSAKCOAFBFODP-ZQOBQRRWSA-N
1153.002
1135.01
≥98.5% (HPLC)
Safety and Handling

Safety and Handling

TSCA : TSCA