missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

α-Cyclodextrin hydrate, 98+%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific AC227291000

 View more versions of this product

Catalog No. AC227291000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chelation/Complexation reagent
Specifications

Specifications

α-Cyclodextrin hydrate
98.0
White
11% max.
Authentic
MFCD00150804
14, 2718
100g
UQIQLAUJPPOBJR-UHFFFAOYNA-N
+148.00 (20.00°C c=2,H2O)
+ 148
60161466
Crystalline Powder
98+%
51211-51-9
100.0
278°C
0.5% max. Linear oligomers (HPLC)
C36H62O31
Glass bottle
972.86 anhydrous, ^a-cyclodextrin hydrate, alpha-cyclodextrin hydrate, alpha-cyclodextrin water, cyclohexaamylose hydrate, water.beta-cyclodextrin
Solubility in water: 145g/L (25°C)
O.OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC1C(O)C2O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
5,10,15,20,25,30-hexakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29-dodecaoxaheptacyclo[26.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶]dotetracontane-31,32,33,34,35,36,37,38,39,40,41,42-dodecol hydrate
990.86
972.85
0.1% max.

Chemical Identifiers

51211-51-9
990.86
UQIQLAUJPPOBJR-UHFFFAOYNA-N
60161466
O.OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC1C(O)C2O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
C36H62O31
MFCD00150804
972.86 anhydrous, ^a-cyclodextrin hydrate, alpha-cyclodextrin hydrate, alpha-cyclodextrin water, cyclohexaamylose hydrate, water.beta-cyclodextrin
5,10,15,20,25,30-hexakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29-dodecaoxaheptacyclo[26.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶]dotetracontane-31,32,33,34,35,36,37,38,39,40,41,42-dodecol hydrate
SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only