Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclobutyl phenyl ketone, 97%

$38.73 - $373.68

Chemical Identifiers

Chemical Name or Material Cyclobutyl phenyl ketone
CAS 5407-98-7
Molecular Formula C11H12O
Formula Weight 160.22
MDL Number MFCD00001326
Beilstein 2044307
Synonym Benzoylcyclobutane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2514603

  SDS
alfa aesar™
B2514603
1g Each for $38.73

AAB2514606

  SDS
alfa aesar™
B2514606
5g Each for $95.99

AAB2514614

  SDS
alfa aesar™
B2514614
25g Each for $373.68
Description & Specifications

Specifications

Formula Weight 160.22
Chemical Name or Material Cyclobutyl phenyl ketone
Assay 97%
Molecular Formula C11H12O
MDL Number MFCD00001326
CAS 5407-98-7
Beilstein 2044307
Synonym Benzoylcyclobutane
Flash Point >110°C (230°F)
Boiling Point 114° to 118°C (7mmHg)
Quantity 1g