Please login to your online account to display your discounted pricing

Cyclobutyl methyl ketone, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Cyclobutyl methyl ketone
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H10O
Linear Formula C4H7COCH3
Formula Weight 98.14
MDL Number MFCD00037148
Synonym Acetylcyclobutane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC377270050

  SDS
thermo fisher scientific
AC377270050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 98.14
Physical Form Liquid
Color Clear colorless to yellow-greenish
Quantity 5g
Density 0.9020g/mL
Beilstein 07, IV, 37
Refractive Index 1.4310 to 1.4330 (20°C, 589nm)
Chemical Name or Material Cyclobutyl methyl ketone
Health Hazard 1 GHS Signal Word: Warning
Molecular Formula C6H10O
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym Acetylcyclobutane
Linear Formula C4H7COCH3
Assay Percent Range 98%
CAS 3019-25-8
MDL Number MFCD00037148
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Flammable liquid and vapor.