missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclobutanone, +98%, ACROS Organics™

Manufacturer:  Acros Organics AC110930250

 View more versions of this product

Catalog No. AC110930250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Cyclobutanone
10°C
Glass bottle
99.0°C
-50.9°C
1191-95-3
C4H6O
unii-6pf2sh405u, cyclobutyloxy, cyclobutanone, 98+%, cyclobutanon, cylcobutanone, cylobutanone, 3-cyclobutanone, 1-oxocyclobutane, cyclobutanone, pubchem9021
C1CC(=O)C1
70.091
70.09
≥98%
0.9300g/mL
Authentic
1.4205 to 1.4225
Colorless to Yellow
25g
98+%
C4H6(=O)
SHQSVMDWKBRBGB-UHFFFAOYSA-N
cyclobutanone
14496
Liquid

Chemical Identifiers

1191-95-3
70.091
unii-6pf2sh405u, cyclobutyloxy, cyclobutanone, 98+%, cyclobutanon, cylcobutanone, cylobutanone, 3-cyclobutanone, 1-oxocyclobutane, cyclobutanone, pubchem9021
cyclobutanone
C4H6O
SHQSVMDWKBRBGB-UHFFFAOYSA-N
14496
C1CC(=O)C1
Safety and Handling

Safety and Handling

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents