missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclobutanol, +99%, ACROS Organics™

Manufacturer:  Acros Organics AC168750050

 View more versions of this product

Catalog No. AC168750050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Cyclobutanol
21°C
Glass bottle
122.0°C to 123.0°C (733.0 mmHg)
5g
99+%
C4H7OH
06, 4
KTHXBEHDVMTNOH-UHFFFAOYSA-N
cyclobutanol
76218
Liquid
0.9200g/mL
Authentic
1.4350 to 1.4370
Colorless
2919-23-5
C4H8O
MFCD00001318
cyclobutyl alcohol, cyclobutyl hydroxide, hydroxycyclobutane, acmc-1cgim, cyclobutanol, cyclobutanol 1g, ksc206s6t, kthxbehdvmtnoh-uhfffaoysa
C1CC(C1)O
72.107
72.11
≥99%

Chemical Identifiers

2919-23-5
72.107
KTHXBEHDVMTNOH-UHFFFAOYSA-N
76218
C1CC(C1)O
C4H8O
MFCD00001318
cyclobutyl alcohol, cyclobutyl hydroxide, hydroxycyclobutane, acmc-1cgim, cyclobutanol, cyclobutanol 1g, ksc206s6t, kthxbehdvmtnoh-uhfffaoysa
cyclobutanol
Safety and Handling

Safety and Handling

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents