missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclobutanol, +99%, ACROS Organics™

Manufacturer:  Acros Organics 168752500

 View more versions of this product

Catalog No. AC168752500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Cyclobutanol
21°C
Colorless
2919-23-5
Glass bottle
≥99%
C4H8O
MFCD00001318
KTHXBEHDVMTNOH-UHFFFAOYSA-N
cyclobutanol
76218
Liquid
0.9200g/mL
122.0°C to 123.0°C (733.0 mmHg)
250mg
Authentic
1.4350 to 1.4370
99+%
C4H7OH
06, 4
C1CC(C1)O
72.107
72.11

Chemical Identifiers

2919-23-5
72.107
KTHXBEHDVMTNOH-UHFFFAOYSA-N
C4H8O
MFCD00001318
76218
Safety and Handling

Safety and Handling

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents