missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclobutanol, 96%

Manufacturer:  Alfa Aesar™ A1243006

 View more versions of this product

Catalog No. AAA1243006


 

Description

Description

Applications
Has synthetic applications of gold-catalyzed ring expansions. Development of novel type of transition metal-catalyzed cascade reactions utilizing small ring systems and the applications.

Solubility
Partly soluble.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acid chlorides, acid anhydrides.
Specifications

Specifications

Cyclobutanol
123°C to 124°C
2919-23-5
C4H8O
UN1120
Hydroxycyclobutane
C1CC(C1)O
72.107
72.11
21°C (69°F)
5g
96%
MFCD00001318
2035937
KTHXBEHDVMTNOH-UHFFFAOYSA-N
cyclobutanol
76218

Chemical Identifiers

2919-23-5
72.107
KTHXBEHDVMTNOH-UHFFFAOYSA-N
76218
C4H8O
MFCD00001318
Hydroxycyclobutane
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTANOLS

TSCA : TSCA

SDS Documents