Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclobutanecarboxamidine hydrochloride, 95%

$29.36 - $193.11

Chemical Identifiers

Chemical Name or Material Cyclobutanecarboxamidine hydrochloride
CAS 71690-89-6
Molecular Formula C5H10N2·HCl
Formula Weight 134.61g/mol
MDL Number MFCD09757543
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27475MC

 
alfa aesar™
H27475-MC
100mg Each for $29.36

AAH27475ME

 
alfa aesar™
H27475-ME
500mg Each for $193.11
Description & Specifications

Specifications

Melting Point ≈160°C
Quantity 100mg
Chemical Name or Material Cyclobutanecarboxamidine hydrochloride
CAS 71690-89-6
Assay 95%
Molecular Formula C5H10N2·HCl
Formula Weight 134.61g/mol
MDL Number MFCD09757543
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.