Learn More
Cyclobutanecarbonitrile, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1030906
Description
It is used for research or analysis use only, as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used for research or analysis use only, as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents, acids.
Specifications
Cyclobutanecarbonitrile | |
0.868 | |
42°C (107°F) | |
C5H7N | |
MFCD00013735 | |
UN1992 | |
cyanocyclobutane, cyclobutyl cyanide, cyclobutancarbonitrile, cyclobutane carbonitrile, acmc-1afpy, cyclobutanecarbonitrile 5g, cyanocyclobutane; cyclobutyl cyanide | |
GFBLFDSCAKHHGX-UHFFFAOYSA-N | |
cyclobutanecarbonitrile | |
78148 | |
97% |
4426-11-3 | |
144°C to 146°C | |
Recognizable | |
1.4325 | |
5 g | |
2037573 | |
Slightly soluble in water. | |
C1CC(C1)C#N | |
81.12 | |
81.12 |
Chemical Identifiers
4426-11-3 | |
81.12 | |
GFBLFDSCAKHHGX-UHFFFAOYSA-N |
C5H7N | |
MFCD00013735 |
Safety and Handling
GHS H Statement
H331-H226-H302-H312
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c
H226-H302+H312+H332
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 224-606-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only