Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cyclin E Ab-2, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and flow cytometry experiments.

Manufacturer: thermo scientific™ lab vision™  MS870P1ABX

 View more versions of this product

Catalog No. MS870P1ABX

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Applications Flow Cytometry
Applications Immunofluorescence
Applications Western Blotting
Clone HE12
Isotype IgG1
Quantity 100µL
Cross Reactivity Human
Antigen Cyclin E Ab-2
Host Species Mouse
Immunogen Recombinant human cyclin E protein
Format Concentrated
Type Primary
Formulation Ab without BSA/Azide (Frozen)
Classification Monoclonal
Conjugate Unlabeled
Research Discipline Cell Cycle
Concentration 1mg/mL

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and flow cytometry experiments with Thermo Scientific™ Cyclin E Ab-2, Mouse Monoclonal Antibody.

Cyclin E is a regulatory subunit of cdk-2 and controls G1/S transition during the mammalian cell cycle. Multiple isoforms of cyclin E are expressed only in tumors but not in the normal tissues. Qualitative and quantitative alterations in cyclin E protein have been implicated as indicators of worse prognosis in various cancers.

Host Species: Mouse

Clone: HE12

Isotype: IgG1

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: Not determined

Immunogen: Recombinant human cyclin E protein

Molecular Weight: Doublet of 50 kDa and a single band of 42 kDa

Positive Control: SKBR3 or LS174T cells; Breast CA or placenta

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Not suitable)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO