missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cyclin D3 Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ Cyclin D3 Ab-2, Mouse Monoclonal Antibody.

$274.10 - $1,459.94

Specifications

Antigen Cyclin D3 Ab-2
Clone DCS-28.1
Applications Immunoprecipitation,Western Blotting,Immunoprecipitation,Western Blotting
Classification Monoclonal
Format Concentrated
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity  
MS216B Thermo Scientific™ Lab Vision™
MS216B
200μg/mL Biotin 1mL Each for $1,459.94
MS216B0 Thermo Scientific™ Lab Vision™
MS216B0
200μg/mL Biotin 100μL Each for $370.88
MS216B1 Thermo Scientific™ Lab Vision™
MS216B1
200μg/mL Biotin 500μL Each for $988.41
MS216P Thermo Scientific™ Lab Vision™
MS216P
200μg/mL Unlabeled 1mL Each for $1,121.67
MS216P0 Thermo Scientific™ Lab Vision™
MS216P0
200μg/mL Unlabeled 100μL Each for $274.10
MS216P1 Thermo Scientific™ Lab Vision™
MS216P1
200μg/mL Unlabeled 500μL Each for $736.37
MS216P1ABX Thermo Scientific™ Lab Vision™
MS216P1ABX
1mg/mL Unlabeled 100μL Each for $833.45
MS216PABX Thermo Scientific™ Lab Vision™
MS216PABX
1mg/mL Unlabeled 200μL Each for $1,218.73
Specifications

Description

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ Cyclin D3 Ab-2, Mouse Monoclonal Antibody.

Cyclin D3 is a G1 cyclin closely related to cyclin D1 and D2. D3 can phosphorylate pRB when associated with cdk4 and/or cdk6.

Host Species: Mouse

Clone: DCS-28.1

Isotype: IgG1

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Purified human recombinant full-length cyclin D3 protein

Molecular Weight: 34 kDa

Positive Control: Ls174T and PC12 cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native verified)
  • Kinase Assay (Not verified)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Cyclin D3 Ab-2
Immunoprecipitation,Western Blotting,Immunoprecipitation,Western Blotting
Concentrated
RUO
Primary
IgG1
DCS-28.1
Monoclonal
Murine
Cell Cycle
Purified human recombinant full length cyclin D3 protein