Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cyclin D3 Ab-2, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ Cyclin D3 Ab-2, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS216P1ABX

 View more versions of this product

Catalog No. MS216P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Cyclin D3 Ab-2
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Clone DCS-28.1
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rat
Format Concentrated
Host Species Murine
Immunogen Purified human recombinant full length cyclin D3 protein
Isotype IgG1
Quantity 100μL
Regulatory Status RUO
Research Discipline Cell Cycle
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human
Target Species Rat

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ Cyclin D3 Ab-2, Mouse Monoclonal Antibody.

Cyclin D3 is a G1 cyclin closely related to cyclin D1 and D2. D3 can phosphorylate pRB when associated with cdk4 and/or cdk6.

Host Species: Mouse

Clone: DCS-28.1

Isotype: IgG1

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Purified human recombinant full-length cyclin D3 protein

Molecular Weight: 34 kDa

Positive Control: Ls174T and PC12 cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native verified)
  • Kinase Assay (Not verified)

Note:

USA: RUO; Int'l: RUO