Please login to your online account to display your discounted pricing

Thermo Scientific™ Cyclin D2 Ab-4, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, flow cytometry and kinase assay experiments.

$272.92 - $1,163.37

Specifications

Antigen Cyclin D2 Ab-4
Clone DCS-3.1 + DCS-5.2
Applications Flow Cytometry
Applications Immunofluorescence
Applications Western Blotting
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS221P

 
thermo scientific™
MS221P
200μg/mL 1mL Each for $1,101.78

MS221P0

 
thermo scientific™
MS221P0
200μg/mL 100µL Each for $272.92

MS221P1

 
thermo scientific™
MS221P1
200μg/mL 500µL Each for $724.59

MS221P1ABX

 
thermo scientific™
MS221P1ABX
1mg/mL 100µL Each for $806.04

MS221PABX

 
thermo scientific™
MS221PABX
1mg/mL 200µL Each for $1,163.37
Description & Specifications

Specifications

Antigen Cyclin D2 Ab-4
Clone DCS-3.1 + DCS-5.2
Applications Flow Cytometry
Applications Immunofluorescence
Applications Western Blotting
Applications Flow Cytometry
Applications Immunofluorescence
Applications Western Blotting
Applications Flow Cytometry
Applications Immunofluorescence
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Cell Cycle
Type Primary
Immunogen Purified histidine-tagged human recombinant full length cyclin D2 protein
Isotype IgG2a IgG2b

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, flow cytometry and kinase assay experiments with Thermo Scientific™ Cyclin D2 Ab-4, Mouse Monoclonal Antibody.

Cyclin D2 is a G1 cyclin required for G1-phase progression and is a strong candidate for a proto-oncogene. Cyclin D2 can phosphorylate pRB when associated with cdk4 and/or cdk6.

Host Species: Mouse

Clone: DCS-3.1 + DCS-5.2

Isotype: IgG2a (DCS-3.1), IgG2b (DCS-5.2)

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Purified histidine-tagged human recombinant full-length cyclin D2 protein

Molecular Weight: 34 kDa

Positive Control: U-2-OS, C82, RD, Bristol-8 or LoVo36 cells. Note that T47D, ZR75, SKBR3, MCF-7, BT-20, MDA468, HBL100, BT5549, HeLa, Saos2 and HT29 cells are negative for cyclin D2.1.

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Immunofluorescence
  • Flow Cytometry
  • Kinase Assay

Note:

USA: RUO; Int'l: RUO