Please login to your online account to display your discounted pricing

Thermo Scientific™ Cyclin D2 Ab-4, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, flow cytometry and kinase assay experiments.

Manufacturer: thermo scientific™  MS221P1ABX

 View more versions of this product

Catalog No. MS221P1ABX

  • / Each
 

Description & Specifications

Specifications

Format Concentrated
Antigen Cyclin D2 Ab-4
Applications Flow Cytometry
Applications Immunofluorescence
Applications Western Blotting
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Regulatory Status RUO
Clone DCS-3.1 + DCS-5.2
Isotype IgG2a IgG2b
Quantity 100µL
Immunogen Purified histidine-tagged human recombinant full length cyclin D2 protein
Type Primary
Conjugate Unlabeled
Classification Monoclonal
Research Discipline Cell Cycle
Concentration 1mg/mL

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, flow cytometry and kinase assay experiments with Thermo Scientific™ Cyclin D2 Ab-4, Mouse Monoclonal Antibody.

Cyclin D2 is a G1 cyclin required for G1-phase progression and is a strong candidate for a proto-oncogene. Cyclin D2 can phosphorylate pRB when associated with cdk4 and/or cdk6.

Host Species: Mouse

Clone: DCS-3.1 + DCS-5.2

Isotype: IgG2a (DCS-3.1), IgG2b (DCS-5.2)

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Purified histidine-tagged human recombinant full-length cyclin D2 protein

Molecular Weight: 34 kDa

Positive Control: U-2-OS, C82, RD, Bristol-8 or LoVo36 cells. Note that T47D, ZR75, SKBR3, MCF-7, BT-20, MDA468, HBL100, BT5549, HeLa, Saos2 and HT29 cells are negative for cyclin D2.1.

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Immunofluorescence
  • Flow Cytometry
  • Kinase Assay

Note:

USA: RUO; Int'l: RUO