missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cyclin D1/Bcl-1, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation applications.

$292.61 - $1,273.98

Specifications

Antigen Cyclin D1/Bcl-1
Applications Immunoprecipitation,Western Blotting,Immunoprecipitation,Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human,Mouse,Human,Mouse
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Format Quantity Price Quantity  
Catalog Number Mfr. No. Format Quantity Price Quantity  
RB9041P Thermo Scientific™ Lab Vision™
RB9041P
Concentrated 1mL Each for $1,273.98
RB9041P0 Thermo Scientific™ Lab Vision™
RB9041P0
Concentrated 100μL Each for $292.61
RB9041P1 Thermo Scientific™ Lab Vision™
RB9041P1
Concentrated 500μL Each for $804.18
RB9041R7 Thermo Scientific™ Lab Vision™
RB9041R7
Prediluted 7mL Each for $497.55
Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation applications with Thermo Scientific™ Cyclin D1/Bcl-1, Rabbit Polyclonal Antibody.

Cyclin D1 or PRAD-1 or bcl-1 is one of the key cell cycle regulators, and functions in association with cdk4 and / or cdk6 by phosphorylating the Rb protein. It is a putative proto-oncogene overexpressed in a wide variety of human neoplasms including mantle cell lymphomas (MCL).

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide from C-terminus of human cyclin D1

Molecular Weight: 36 kDa

Positive Control: Breast CA or mantle cell lymphoma

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Cyclin D1/Bcl-1
Polyclonal
Human,Mouse,Human,Mouse
RUO
Primary
IgG
Immunoprecipitation,Western Blotting,Immunoprecipitation,Western Blotting
Unlabeled
Rabbit
Cell Cycle
A synthetic peptide from C-terminus of human cyclin D1