Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cyclin D1/Bcl-1 Ab-4, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in western blotting, immunofluorescence and immunoprecipitation experiments.

$274.10 - $1,218.73

Specifications

Antigen Cyclin D1/Bcl-1 Ab-4
Concentration 1mg/mL
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB212P

 
thermo scientific™ lab vision™
RB212P
Purified Ab with BSA and Azide 1mL Each for $1,121.67

RB212P0

 
thermo scientific™ lab vision™
RB212P0
Purified Ab with BSA and Azide 100µL Each for $274.10

RB212P1

 
thermo scientific™ lab vision™
RB212P1
Purified Ab with BSA and Azide 500µL Each for $704.06

RB212P1ABX

 
thermo scientific™ lab vision™
RB212P1ABX
Ab without BSA/Azide (Frozen) 500µL Each for $806.57

RB212PABX

 
thermo scientific™ lab vision™
RB212PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,218.73
Description & Specifications

Specifications

Antigen Cyclin D1/Bcl-1 Ab-4
Concentration 1mg/mL
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Host Species Rabbit
Regulatory Status RUO
Research Discipline Cell Cycle
Type Primary
Immunogen A synthetic peptide derived from the C-terminal of human cyclin D1
Isotype IgG

Ensure accurate, reproducible results in western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ Cyclin D1/Bcl-1 Ab-4, Rabbit Polyclonal Antibody.

Cyclin D1or PRAD-1 or bcl-1 is one of the key cell cycle regulators, and functions in association with cdk4 and/or cdk6 by phosphorylating the Rb protein. It is a putative proto-oncogene overexpressed in a wide variety of human neoplasms including mantle cell lymphomas (MCL).

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: C-terminus

Immunogen: A synthetic peptide derived from the C-terminal of human cyclin D1

Molecular Weight: 36 kDa

Positive Control: MAD109 cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO