Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cyclin D1/Bcl-1 Ab-4, Rabbit Polyclonal Antibody


Ensure accurate, reproducible results in western blotting, immunofluorescence and immunoprecipitation experiments.

Manufacturer: thermo scientific™ lab vision™  RB212P1ABX

 View more versions of this product

Catalog No. RB212P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunoprecipitation
Applications Western Blotting
Quantity 500µL
Isotype IgG
Regulatory Status RUO
Antigen Cyclin D1/Bcl-1 Ab-4
Cross Reactivity Human
Cross Reactivity Mouse
Immunogen A synthetic peptide derived from the C-terminal of human cyclin D1
Format Concentrated
Host Species Rabbit
Classification Polyclonal
Concentration 1mg/mL
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Cell Cycle
Type Primary
Conjugate Unlabeled
Item Description 500uL; 1mg/mL; Unlabeled; Purified without BSA and azide

Ensure accurate, reproducible results in western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ Cyclin D1/Bcl-1 Ab-4, Rabbit Polyclonal Antibody.

Cyclin D1or PRAD-1 or bcl-1 is one of the key cell cycle regulators, and functions in association with cdk4 and/or cdk6 by phosphorylating the Rb protein. It is a putative proto-oncogene overexpressed in a wide variety of human neoplasms including mantle cell lymphomas (MCL).

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: C-terminus

Immunogen: A synthetic peptide derived from the C-terminal of human cyclin D1

Molecular Weight: 36 kDa

Positive Control: MAD109 cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO