missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cyclin D1/Bcl-1 Ab-3, Rabbit Polyclonal Antibody

Obtain accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation and kinase assay application.

$713.82 - $1,195.12

Specifications

Antigen Cyclin D1/Bcl-1 Ab-3
Concentration 1mg/mL
Applications Immunoprecipitation,Western Blotting,Immunoprecipitation,Western Blotting
Classification Polyclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
RB010P Thermo Scientific™ Lab Vision™
RB010P
Purified Ab with BSA and Azide 1mL Each for $1,140.29
RB010P1 Thermo Scientific™ Lab Vision™
RB010P1
Purified Ab with BSA and Azide 500μL Each for $713.82
RB010P1ABX Thermo Scientific™ Lab Vision™
RB010P1ABX
Ab without BSA/Azide (Frozen) 500μL Each for $783.39
RB010PABX Thermo Scientific™ Lab Vision™
RB010PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,195.12
Specifications

Description

Obtain accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation and kinase assay applications with Thermo Scientific™ Cyclin D1/Bcl-1 Ab-3, Rabbit Polyclonal Antibody.

Cyclin D1 (also known as PRAD-1 or bcl-1), one of the key cell cycle regulators, is a putative proto-oncogene overexpressed in a wide variety of human neoplasms. About 50-70% of mantle cell lymphomas (MCL) contain a t (11; 14)(q13; q32) translocation resulting in over-expression of cyclin D1.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from the C-terminus of human cyclin D1

Molecular Weight: 36 kDa

Positive Control: MAD109 cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Kinase Assay

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Cyclin D1/Bcl-1 Ab-3
Immunoprecipitation,Western Blotting,Immunoprecipitation,Western Blotting
Unlabeled
Concentrated
RUO
Primary
IgG
1mg/mL
Polyclonal
Human,Mouse,Human,Mouse
Rabbit
Cell Cycle
A synthetic peptide derived from the C-terminus of human cyclin D1