Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cyclin D1/Bcl-1 Ab-3, Rabbit Polyclonal Antibody


Obtain accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation and kinase assay application.

Manufacturer: thermo scientific™ lab vision™  RB010P1ABX

 View more versions of this product

Catalog No. RB010P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Cyclin D1/Bcl-1 Ab-3
Applications Immunoprecipitation
Applications Western Blotting
Quantity 500µL
Isotype IgG
Cross Reactivity Human
Cross Reactivity Mouse
Immunogen A synthetic peptide derived from the C-terminus of human cyclin D1
Format Concentrated
Regulatory Status RUO
Host Species Rabbit
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Cell Cycle
Type Primary
Classification Polyclonal
Conjugate Unlabeled
Concentration 1mg/mL
Item Description 500uL; Purified without BSA and azide

Obtain accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation and kinase assay applications with Thermo Scientific™ Cyclin D1/Bcl-1 Ab-3, Rabbit Polyclonal Antibody.

Cyclin D1 (also known as PRAD-1 or bcl-1), one of the key cell cycle regulators, is a putative proto-oncogene overexpressed in a wide variety of human neoplasms. About 50-70% of mantle cell lymphomas (MCL) contain a t (11; 14)(q13; q32) translocation resulting in over-expression of cyclin D1.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from the C-terminus of human cyclin D1

Molecular Weight: 36 kDa

Positive Control: MAD109 cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Kinase Assay

Note:

USA: RUO; Int'l: RUO