missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Cyclin D1/Bcl-1 Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in kinase assay applications with Thermo Scientific™ Cyclin D1/Bcl-1 Ab-2, Mouse Monoclonal Antibody.

$274.10 - $1,217.59

Specifications

Antigen Cyclin D1/Bcl-1 Ab-2
Clone DCS-11
Applications Kinase Assay
Cross Reactivity Human,Mouse,Rat,Human,Mouse,Rat,Human,Mouse,Rat
Format Purified
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS211P Thermo Scientific™
MS211P
200μg/mL 1mL Each for $1,120.63
MS211P0 Thermo Scientific™
MS211P0
200μg/mL 100μL Each for $274.10
MS211P1 Thermo Scientific™
MS211P1
200μg/mL 500μL Each for $743.69
MS211P1ABX Thermo Scientific™
MS211P1ABX
1mg/mL 100μL Each for $832.69
MS211PABX Thermo Scientific™
MS211PABX
1mg/mL 200μL Each for $1,217.59
Specifications

Description

Ensure accurate, reproducible results in kinase assay applications with Thermo Scientific™ Cyclin D1/Bcl-1 Ab-2, Mouse Monoclonal Antibody.

Cyclin D1or PRAD-1 or bcl-1 is one of the key cell cycle regulators, and functions in association with cdk4 and/or cdk6 by phosphorylating the Rb protein. It is a putative proto-oncogene overexpressed in a wide variety of human neoplasms including mantle cell lymphomas (MCL).

Host Species: Mouse

Clone: DCS-11

Isotype: IgG2a

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Human recombinant full-length cyclin D1 protein

Molecular Weight: 36 kDa

Cellular Localization: Nuclear

Recommended for:

  • Kinase Assay

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Cyclin D1/Bcl-1 Ab-2
Kinase Assay
Purified
Primary
IgG2a
DCS-11
Human,Mouse,Rat,Human,Mouse,Rat,Human,Mouse,Rat
Murine
Human recombinant full length cyclin D1 protein