Please login to your online account to display your discounted pricing

Thermo Scientific™ Cyclin D1/Bcl-1 Ab-2, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in kinase assay applications with Thermo Scientific™ Cyclin D1/Bcl-1 Ab-2, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™  MS211P1ABX

 View more versions of this product

Catalog No. MS211P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Kinase Assay
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Immunogen Human recombinant full length cyclin D1 protein
Host Species Mouse
Format Purified
Antigen Cyclin D1/Bcl-1 Ab-2
Quantity 100µL
Isotype IgG2a
Clone DCS-11
Type Primary
Concentration 1mg/mL

Ensure accurate, reproducible results in kinase assay applications with Thermo Scientific™ Cyclin D1/Bcl-1 Ab-2, Mouse Monoclonal Antibody.

Cyclin D1or PRAD-1 or bcl-1 is one of the key cell cycle regulators, and functions in association with cdk4 and/or cdk6 by phosphorylating the Rb protein. It is a putative proto-oncogene overexpressed in a wide variety of human neoplasms including mantle cell lymphomas (MCL).

Host Species: Mouse

Clone: DCS-11

Isotype: IgG2a

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Human recombinant full-length cyclin D1 protein

Molecular Weight: 36 kDa

Cellular Localization: Nuclear

Recommended for:

  • Kinase Assay

Note:

USA: RUO; Int'l: RUO