missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Epredia™ Lab Vision™ Cyclin D1/Bcl-1 Ab-1 Mouse Monoclonal Antibody, 1mL; 200μg/mL, biotinylated, BSA and azide
Ensure accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation, biological blockade and flow cytometry applications.
$945.10 - $945.10
Specifications
Antigen | Cyclin D1/Bcl-1 Ab-1 |
---|---|
Clone | DCS-6 |
Concentration | 200μg/mL |
Applications | Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Conjugate | Concentration | Quantity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Conjugate | Concentration | Quantity | Price | Quantity | |||
MS210P1
|
Epredia™
MS210P1 |
Unconjugated | 200μg/mL | 500 μL |
Each for $945.10
|
|
|||
Description
Cyclin D1or PRAD-1 or bcl-1 is one of the key cell cycle regulators and is a putative proto-oncogene overexpressed in a wide variety of human neoplasms including mantle cell lymphomas (MCL)Host Species: Mouse
Clone: DCS-6
Isotype: IgG2a/κ
Species Reactivity: Human, Monkey, Mouse, Rat and Dog.
Epitope: Not determined
Immunogen: Human recombinant full-length cyclin D1 protein
Molecular Weight: 36 kDa
Positive Control: MAD109 cells; Mantle cell lymphomas and breast CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Not Suitable)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
- Biological Blockade
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
Cyclin D1/Bcl-1 Ab-1 | |
200μg/mL | |
Monoclonal | |
Concentrated | |
RUO | |
Canine, Human, Mouse, Non-human Primate, Rat | |
IgG2a κ |
DCS-6 | |
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Cell Cycle | |
Human recombinant full length cyclin D1 protein | |
Primary |