Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cyclin D1/Bcl-1 Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation, biological blockade and flow cytometry applications.

Manufacturer: thermo scientific™ lab vision™  MS210P1ABX

 View more versions of this product

Catalog No. MS210P1ABX

  • / Each
 

Description & Specifications

Specifications

Quantity 100µL
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Format Concentrated
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Cross Reactivity Rat
Host Species Mouse
Antigen Cyclin D1/Bcl-1 Ab-1
Isotype IgG2a kappa
Regulatory Status RUO
Clone DCS-6
Immunogen Human recombinant full length cyclin D1 protein
Type Primary
Classification Monoclonal
Research Discipline Cell Cycle
Concentration 1mg/mL
Conjugate Unlabeled

Ensure accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation, biological blockade and flow cytometry applications with Thermo Scientific™ Cyclin D1/Bcl-1 Ab-1, Mouse Monoclonal Antibody.

Cyclin D1or PRAD-1 or bcl-1 is one of the key cell cycle regulators and is a putative proto-oncogene overexpressed in a wide variety of human neoplasms including mantle cell lymphomas (MCL)

Host Species: Mouse

Clone: DCS-6

Isotype: IgG2a/κ

Species Reactivity: Human, Monkey, Mouse, Rat and Dog.

Epitope: Not determined

Immunogen: Human recombinant full-length cyclin D1 protein

Molecular Weight: 36 kDa

Positive Control: MAD109 cells; Mantle cell lymphomas and breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Not Suitable)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Biological Blockade
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO