Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cyclin C Ab-1, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation and kinase assay applications.

$244.18 - $1,217.59

Specifications

Antigen Cyclin C Ab-1
Concentration 1mg/mL
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB009P

 
thermo scientific™ lab vision™
RB009P
Purified Ab with BSA and Azide 1mL Each for $1,131.65

RB009P0

 
thermo scientific™ lab vision™
RB009P0
Purified Ab with BSA and Azide 100µL Each for $244.18

RB009P1

 
thermo scientific™ lab vision™
RB009P1
Purified Ab with BSA and Azide 500µL Each for $703.41

RB009P1ABX

 
thermo scientific™ lab vision™
RB009P1ABX
Ab without BSA/Azide (Frozen) 500µL Each for $797.16

RB009PABX

 
thermo scientific™ lab vision™
RB009PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,217.59
Description & Specifications

Specifications

Antigen Cyclin C Ab-1
Concentration 1mg/mL
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Host Species Rabbit
Regulatory Status RUO
Research Discipline Cell Cycle
Type Primary
Immunogen A synthetic peptide corresponding to aa 290-303 (Cys-QGPNGSQNSSYSQS) from the C-terminal of human cyclin C
Isotype IgG

Ensure accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation and kinase assay applications with Thermo Scientific™ Cyclin C Ab-1, Rabbit Polyclonal Antibody.

Cyclin C was originally identified by a genetic screen for human and Drosophila cDNAs that complement a triple knock-out of the CLN genes in Saccharomyces cerevisiae. Cyclin C is a nuclear protein present in a multiprotein complex. It interacts both in vitro and in vivo with cdk8, a novel protein-kinase of the cdk family. It is suggested that cyclin C and cdk8 control RNA polymerase II function. Human cdk8-cyclin C might be functionally associated with the mammalian transcription apparatus, perhaps involved in relaying growth-regulatory signals.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Dog, Mouse and Rat. Others not known.

Epitope: aa 290-303

Immunogen: A synthetic peptide corresponding to aa 290-303 (Cys-QGPNGSQNSSYSQS) from the C-terminal of human cyclin C

Molecular Weight: 31-32 kDa

Positive Control: HeLa cells

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation
  • Kinase Assay

Note:

USA: RUO; Int'l: RUO