Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cyclin C Ab-1, Rabbit Polyclonal Antibody


Ensure accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation and kinase assay applications.

Manufacturer: thermo scientific™ lab vision™  RB009P1ABX

 View more versions of this product

Catalog No. RB009P1ABX

  • / Each
 

Description & Specifications

Specifications

Immunogen A synthetic peptide corresponding to aa 290-303 (Cys-QGPNGSQNSSYSQS) from the C-terminal of human cyclin C
Host Species Rabbit
Antigen Cyclin C Ab-1
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Quantity 500µL
Isotype IgG
Format Concentrated
Conjugate Unlabeled
Formulation Ab without BSA/Azide (Frozen)
Concentration 1mg/mL
Research Discipline Cell Cycle
Classification Polyclonal
Type Primary

Ensure accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation and kinase assay applications with Thermo Scientific™ Cyclin C Ab-1, Rabbit Polyclonal Antibody.

Cyclin C was originally identified by a genetic screen for human and Drosophila cDNAs that complement a triple knock-out of the CLN genes in Saccharomyces cerevisiae. Cyclin C is a nuclear protein present in a multiprotein complex. It interacts both in vitro and in vivo with cdk8, a novel protein-kinase of the cdk family. It is suggested that cyclin C and cdk8 control RNA polymerase II function. Human cdk8-cyclin C might be functionally associated with the mammalian transcription apparatus, perhaps involved in relaying growth-regulatory signals.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Dog, Mouse and Rat. Others not known.

Epitope: aa 290-303

Immunogen: A synthetic peptide corresponding to aa 290-303 (Cys-QGPNGSQNSSYSQS) from the C-terminal of human cyclin C

Molecular Weight: 31-32 kDa

Positive Control: HeLa cells

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation
  • Kinase Assay

Note:

USA: RUO; Int'l: RUO