Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cyclin B1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry, western blotting and flow cytometry applications with Thermo Scientific™ Cyclin B1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS338P1ABX

 View more versions of this product

Catalog No. MS338P1ABX

  • / Each
 

Description & Specifications

Specifications

Immunogen Hamster cyclin B1 protein
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Clone V152
Quantity 100µL
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen Cyclin B1
Regulatory Status RUO
Host Species Mouse
Format Concentrated
Isotype IgG1
Conjugate Unlabeled
Classification Monoclonal
Research Discipline Cell Cycle
Type Primary
Formulation Ab without BSA/Azide (Frozen)
Concentration 1mg/mL

Ensure accurate, reproducible results in immunohistochemistry, western blotting and flow cytometry applications with Thermo Scientific™ Cyclin B1, Mouse Monoclonal Antibody.

In mammals, cyclin B associates with inactive p34cdc2. Cyclin Bp34cdc2 plays a critical role in G2 to M transition.

Host Species: Mouse

Clone: V152

Isotype: IgG1

Species Reactivity: Human, Mouse, Rat and Hamster. Others not known.

Epitope: Not determined

Immunogen: Hamster cyclin B1 protein

Molecular Weight: 62 kDa

Positive Control: Jurkat cells, Tonsil or breast CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Not suitable)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO