Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cyclin B1 Ab-4, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence and immunoprecipitation experiments.

Manufacturer: thermo scientific™ lab vision™  MS869P1ABX

 View more versions of this product

Catalog No. MS869P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Cyclin B1 Ab-4
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Classification Monoclonal
Clone GNS11
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Murine
Immunogen Human cyclin B1 protein
Isotype IgG2a
Quantity 100μL
Regulatory Status RUO
Research Discipline Cell Cycle
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Hamster
Target Species Human
Target Species Murine
Target Species Rat

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ Cyclin B1 Ab-4, Mouse Monoclonal Antibody.

In mammals, cyclin B associates with inactive p34cdc2. Cyclin B-p34cdc2 plays a critical role in G2 to M transition.

Host Species: Mouse

Clone: GNS11

Isotype: IgG2a

Species Reactivity: Human, Mouse, Rat and Hamster. Others not known.

Epitope: aa 1-306

Immunogen: Human cyclin B1 protein

Molecular Weight: 62 kDa

Positive Control: LS174T cells, Tonsil or breast CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO