Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cyclin B1 Ab-3, Mouse Monoclonal Antibody

Obtain accurate and reproducible results in western blotting, immunofluorescence, immunoprecipitation and flow cytometry experiments.

$274.55 - $1,245.41

Specifications

Antigen Cyclin B1 Ab-3
Clone GNS1
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS868P

 
thermo scientific™ lab vision™
MS868P
200μg/mL 1mL Each for $1,169.39

MS868P0

 
thermo scientific™ lab vision™
MS868P0
200μg/mL 100μL Each for $274.55

MS868P1

 
thermo scientific™ lab vision™
MS868P1
200μg/mL 500μL Each for $769.65

MS868P1ABX

 
thermo scientific™ lab vision™
MS868P1ABX
1mg/mL 100μL Each for $854.67

MS868PABX

 
thermo scientific™ lab vision™
MS868PABX
1mg/mL 200μL Each for $1,245.41
Description & Specifications

Specifications

Antigen Cyclin B1 Ab-3
Clone GNS1
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Cell Cycle
Type Primary
Immunogen Human cyclin B1 protein
Isotype IgG1

Obtain accurate and reproducible results in western blotting, immunofluorescence, immunoprecipitation and flow cytometry experiments with Thermo Scientific™ Cyclin B1 Ab-3, Mouse Monoclonal Antibody.

In mammals, cyclin B associates with inactive p34cdc2. Cyclin B-p34cdc2 plays a critical role in G2 to M transition.

Host Species: Mouse

Clone: GNS1

Isotype: IgG1

Species Reactivity: Human, Mouse and Hamster. Others not known.

Epitope: aa 1-21

Immunogen: Human cyclin B1 protein

Molecular Weight: 62 kDa

Positive Control: MAD109 or LS174T cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO