Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cyclin B1 Ab-3, Mouse Monoclonal Antibody


Obtain accurate and reproducible results in western blotting, immunofluorescence, immunoprecipitation and flow cytometry experiments.

Manufacturer: thermo scientific™ lab vision™  MS868P1ABX

 View more versions of this product

Catalog No. MS868P1ABX

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Antigen Cyclin B1 Ab-3
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Quantity 100µL
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Immunogen Human cyclin B1 protein
Isotype IgG1
Format Concentrated
Regulatory Status RUO
Clone GNS1
Conjugate Unlabeled
Type Primary
Research Discipline Cell Cycle
Concentration 1mg/mL
Classification Monoclonal

Obtain accurate and reproducible results in western blotting, immunofluorescence, immunoprecipitation and flow cytometry experiments with Thermo Scientific™ Cyclin B1 Ab-3, Mouse Monoclonal Antibody.

In mammals, cyclin B associates with inactive p34cdc2. Cyclin B-p34cdc2 plays a critical role in G2 to M transition.

Host Species: Mouse

Clone: GNS1

Isotype: IgG1

Species Reactivity: Human, Mouse and Hamster. Others not known.

Epitope: aa 1-21

Immunogen: Human cyclin B1 protein

Molecular Weight: 62 kDa

Positive Control: MAD109 or LS174T cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO