missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cyclin A Ab-7, Rabbit Polyclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments. 

$277.39 - $1,411.39

Specifications

Antigen Cyclin A Ab-7
Concentration 1mg/mL
Applications Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blotting,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blotting,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blotting
Classification Polyclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
RB1548P Thermo Scientific™ Lab Vision™
RB1548P
Purified Ab with BSA and Azide 1mL Each for $1,284.94
RB1548P0 Thermo Scientific™ Lab Vision™
RB1548P0
Purified Ab with BSA and Azide 100μL Each for $277.39
RB1548P1 Thermo Scientific™ Lab Vision™
RB1548P1
Purified Ab with BSA and Azide 500μL Each for $769.37
RB1548P1ABX Thermo Scientific™ Lab Vision™
RB1548P1ABX
Ab without BSA/Azide (Frozen) 500μL Each for $863.51
RB1548PABX Thermo Scientific™ Lab Vision™
RB1548PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,411.39
Specifications

Description

Obtain accurate, reproducible results in Immunohistochemistry, Western Blotting and Immunoprecipitation experiments with Thermo Scientific™ Cyclin A Ab-7, Rabbit Polyclonal Antibody.

Cyclins are regulatory subunits of the cyclin-dependent kinases (cdk's). They are expressed temporally to control transition at different specific phases of the cell cycle. In mammalian somatic cells, cyclin A is required for S-phase and passage through G2-phase.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide from the C-terminus of human cyclin A

Molecular Weight: 60 kDa

Positive Control: HeLa cells, Tonsil

Cellular Localization: Nuclear and occasional cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Cyclin A Ab-7
Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blotting,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blotting,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blotting
Unlabeled
Concentrated
RUO
Primary
IgG
1mg/mL
Polyclonal
Human,Mouse,Rat,Human,Mouse,Rat,Human,Mouse,Rat
Rabbit
Cell Cycle
A synthetic peptide from the C-terminus of human cyclin A