Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cyclin A Ab-7, Rabbit Polyclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments. 

Manufacturer: thermo scientific™ lab vision™  RB1548PABX

 View more versions of this product

Catalog No. RB1548PABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen Cyclin A Ab-7
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Immunogen A synthetic peptide from the C-terminus of human cyclin A
Quantity 1mL
Format Concentrated
Host Species Rabbit
Regulatory Status RUO
Isotype IgG
Type Primary
Conjugate Unlabeled
Research Discipline Cell Cycle
Concentration 1mg/mL
Classification Polyclonal
Formulation Ab without BSA/Azide (Frozen)

Obtain accurate, reproducible results in Immunohistochemistry, Western Blotting and Immunoprecipitation experiments with Thermo Scientific™ Cyclin A Ab-7, Rabbit Polyclonal Antibody.

Cyclins are regulatory subunits of the cyclin-dependent kinases (cdk's). They are expressed temporally to control transition at different specific phases of the cell cycle. In mammalian somatic cells, cyclin A is required for S-phase and passage through G2-phase.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide from the C-terminus of human cyclin A

Molecular Weight: 60 kDa

Positive Control: HeLa cells, Tonsil

Cellular Localization: Nuclear and occasional cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO