missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclic Pifithrin-alpha Hydrobromide, TCI America™

Manufacturer:  TCI America C282620MG

 View more versions of this product

Catalog No. C282620MG



Specifications

Specifications

Cyclic Pifithrin-alpha Hydrobromide
20mg
C16H17BrN2S
pifithrin-beta hydrobromide, cyclic-pifithrin-alpha, cyclic pifithrin-alpha hydrobromide, pifithrin-alpha, cyclic, unii-up003xxt4d, up003xxt4d, 2-4-methylphenyl imidazo 2,1-b-5,6,7,8-tetrahydrobenzothiazole, hbr, 2-4-methylphenyl imidazo 2,1-b-5,6,7,8-tetrahydrobenzothiazole hydrobromide, cyclic pifithrin-, a hydrobromide, pifithrin-beta hydrobromide mi
CC1=CC=C(C=C1)C2=CN3C4=C(CCCC4)SC3=N2.Br
349.29
349.29
≥98.0% (HPLC)
White
511296-88-1
MFCD02683960
SGNCOAOESGSEOP-UHFFFAOYSA-N
2-(4-methylphenyl)-5,6,7,8-tetrahydroimidazo[2,1-b][1,3]benzothiazole;hydrobromide
11515812
Crystal-Powder at 20°C

Chemical Identifiers

511296-88-1
349.29
SGNCOAOESGSEOP-UHFFFAOYSA-N
11515812
CC1=CC=C(C=C1)C2=CN3C4=C(CCCC4)SC3=N2.Br
C16H17BrN2S
MFCD02683960
pifithrin-beta hydrobromide, cyclic-pifithrin-alpha, cyclic pifithrin-alpha hydrobromide, pifithrin-alpha, cyclic, unii-up003xxt4d, up003xxt4d, 2-4-methylphenyl imidazo 2,1-b-5,6,7,8-tetrahydrobenzothiazole, hbr, 2-4-methylphenyl imidazo 2,1-b-5,6,7,8-tetrahydrobenzothiazole hydrobromide, cyclic pifithrin-, a hydrobromide, pifithrin-beta hydrobromide mi
2-(4-methylphenyl)-5,6,7,8-tetrahydroimidazo[2,1-b][1,3]benzothiazole;hydrobromide
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

Documents