Learn More
Cyclen, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 294320500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagentSpecifications
Cyclen | |
294-90-6 | |
White to Yellow | |
97% | |
C8H20N4 | |
50 mg | |
cyclen, 1,4,7,10-tetraazacyclododecane, 12 anen4, 1,4,7,10tetraaza-cyclododecane, 5-26-11-00023 beilstein handbook reference, 1,4,7,10-tetracyclododecane, 1,4,7,10-tetraazacyclododecane cyclen, tetraaza-12-crown-4, pubchem23801 | |
QBPPRVHXOZRESW-UHFFFAOYSA-N | |
1,4,7,10-tetrazacyclododecane | |
64963 | |
172.27 | |
Crystalline Powder |
97% | |
108.0°C to 113.0°C | |
Authentic | |
Glass bottle | |
MFCD00066281 | |
15, 2699 | |
Solubility in water: soluble | |
C1CNCCNCCNCCN1 | |
172.27 | |
CHEBI:37391 | |
97% |
Chemical Identifiers
294-90-6 | |
172.27 | |
QBPPRVHXOZRESW-UHFFFAOYSA-N | |
64963 | |
1,4,7,10-tetrazacyclododecane |
C8H20N4 | |
MFCD00066281 | |
cyclen, 1,4,7,10-tetraazacyclododecane, 12 anen4, 1,4,7,10tetraaza-cyclododecane, 5-26-11-00023 beilstein handbook reference, 1,4,7,10-tetracyclododecane, 1,4,7,10-tetraazacyclododecane cyclen, tetraaza-12-crown-4, pubchem23801 | |
CHEBI:37391 | |
C1CNCCNCCNCCN1 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im
GHS Signal Word: Danger
RTECSNumber : XA5253000
RUO – Research Use Only