missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclen, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 294322500

 View more versions of this product

Catalog No. AC294322500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Chelation/Complexation reagent
Specifications

Specifications

Cyclen
Glass bottle
108.0°C to 113.0°C
97%
97%
MFCD00066281
1,4,7,10-tetraazacyclododecane, 1,4,7,10-tetraazacyclododecane cyclen, 1,4,7,10-tetracyclododecane, 1,4,7,10tetraaza-cyclododecane, 12 anen4, 5-26-11-00023 beilstein handbook reference, cyclen, pubchem23801, tetraaza-12-crown-4
QBPPRVHXOZRESW-UHFFFAOYSA-N
1,4,7,10-tetrazacyclododecane
64963
172.27
97%
Authentic
White to Yellow
250mg
294-90-6
C8H20N4
15, 2699
Solubility in water: soluble
C1CNCCNCCNCCN1
172.27
CHEBI:37391
Crystalline Powder

Chemical Identifiers

294-90-6
172.27
QBPPRVHXOZRESW-UHFFFAOYSA-N
64963
1,4,7,10-tetrazacyclododecane
C8H20N4
MFCD00066281
1,4,7,10-tetraazacyclododecane, 1,4,7,10-tetraazacyclododecane cyclen, 1,4,7,10-tetracyclododecane, 1,4,7,10tetraaza-cyclododecane, 12 anen4, 5-26-11-00023 beilstein handbook reference, cyclen, pubchem23801, tetraaza-12-crown-4
CHEBI:37391
C1CNCCNCCNCCN1
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im

GHS Signal Word: Danger

RTECSNumber : XA5253000

SDS Documents