Please login to your online account to display your discounted pricing

Cyclapolin 9, Tocris

Selective, ATP-competitive PLK1 inhibitor

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material Cyclapolin 9
Assay Percent Range >98%
Molecular Formula C9H4F3N3O4S
Formula Weight 307.21
Synonym 7-Nitro-5-(trifluoromethyl)-2-benzothiazolecarboxamide-3-oxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

311610

 
r & d systems
RDS311610
10mg Each for $181.19

311650

 
r & d systems
RDS311650
50mg Each for $762.58
Description & Specifications

Specifications

Formula Weight 307.21
Quantity 10mg
Chemical Name or Material Cyclapolin 9
Assay Percent Range >98%
Molecular Formula C9H4F3N3O4S
CAS 40533-25-3
Synonym 7-Nitro-5-(trifluoromethyl)-2-benzothiazolecarboxamide-3-oxide
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C