missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyanuric fluoride, 98%

Manufacturer:  Alfa Aesar™ A1566606

 View more versions of this product

Catalog No. AAA1566606



Description

Description

Applications
Cyanuric fluoride acts as a fluorinating agent used in the conversion of carboxylic acids into acyl fluorides. It is used as a precursor for fibre-reactive dyes. It is a specific reagent for tyrosine residues in enzyme. Further, it is involved in the preparation of cyanuric acid by hydrolysis

Solubility
Miscible with chloroform, carbon tetrachloride,ether, dioxane and ketones.

Notes
May precipitate upon storage. Moisture sensitive. Incompatible strong oxidizing agents, strong bases and alcohols.
Specifications

Specifications

Cyanuric fluoride
73°C to 74°C
5g
C3F3N3
UN3389
cyanuric fluoride, trifluorotriazine, cyanuric trifluoride, trifluoro-s-triazine, 1,3,5-triazine, 2,4,6-trifluoro, 2,4,6-trifluoro-s-triazine, s-triazine, 2,4,6-trifluoro, cyanuryl fluoride, unii-31kw4s5fbb, s-triazine, 2,4,6-trifluro
C1(=NC(=NC(=N1)F)F)F
135.049
135.05
Acrid
-38°C
675-14-9
MFCD00014597
124237
VMKJWLXVLHBJNK-UHFFFAOYSA-N
2,4,6-trifluoro-1,3,5-triazine
12664
98%

Chemical Identifiers

675-14-9
135.049
VMKJWLXVLHBJNK-UHFFFAOYSA-N
12664
C1(=NC(=NC(=N1)F)F)F
C3F3N3
MFCD00014597
cyanuric fluoride, trifluorotriazine, cyanuric trifluoride, trifluoro-s-triazine, 1,3,5-triazine, 2,4,6-trifluoro, 2,4,6-trifluoro-s-triazine, s-triazine, 2,4,6-trifluoro, cyanuryl fluoride, unii-31kw4s5fbb, s-triazine, 2,4,6-trifluro
2,4,6-trifluoro-1,3,5-triazine
Safety and Handling

Safety and Handling

GHS H Statement
H310-H330-H314-H318
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P304+P340-P305+P351+P338-P320-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, CORROSIVE, N.O.S.

RTECSNumber : XZ1750000

TSCA : TSCA

Documents