missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyanuric Acid, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1544722

 View more versions of this product

Catalog No. AAA1544722



Description

Description

Applications
Cyanuric acid acts as a chlorine stabilizer in swimming pools. It is used as a precursor to N-chlorinated cyanurates, which are used to disinfect water and crosslinking agents for polyurethane resins. It serves as a process regulator. Further, it is used in the diagnostic determination of melamine and related compounds in kidney tissue.

Solubility
Soluble in sulfuric acid, dimethylformamide, sodium hydroxide, potassium hydroxide and diemthyl sulfoxide. Slightly soluble in acetone, benzene, diethyl ether, ethanol and hexane. Insoluble in cold methanol, benzene and chloroform.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Cyanuric acid
99%
MFCD00082990
14,2698
ZFSLODLOARCGLH-UHFFFAOYSA-N
1,3,5-triazinane-2,4,6-trione
7956
129.08
>360°C
108-80-5
C3H3N3O3
126982
cyanuric acid, isocyanuric acid, 1,3,5-triazine-2,4,6-triol, trihydroxycyanidine, tricyanic acid, sym-triazinetriol, s-triazinetriol, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, pseudocyanuric acid, isocyanurate acid
C1(=O)NC(=O)NC(=O)N1
129.075
CHEBI:38028
99%
100g

Chemical Identifiers

108-80-5
129.075
ZFSLODLOARCGLH-UHFFFAOYSA-N
7956
1,3,5-triazinane-2,4,6-trione
C3H3N3O3
MFCD00082990
cyanuric acid, isocyanuric acid, 1,3,5-triazine-2,4,6-triol, trihydroxycyanidine, tricyanic acid, sym-triazinetriol, s-triazinetriol, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, pseudocyanuric acid, isocyanurate acid
CHEBI:38028
C1(=O)NC(=O)NC(=O)N1
Safety and Handling

Safety and Handling

RTECSNumber : XZ1800000

TSCA : TSCA

storageNote1 : Hygroscopic

Documents