Please login to your online account to display your discounted pricing

Cyanopropylmethyldichlorosilane, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Cyanopropylmethyldichlorosilane
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H9Cl2NSi
Formula Weight 182.13
Synonym Dichloro(3-cyanopropyl)methylsilane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC432250050

  SDS
thermo fisher scientific
AC432250050
Glass bottle 5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC432250250

  SDS
thermo fisher scientific
AC432250250
Glass bottle 25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥96.0%
Formula Weight 182.13
Quantity 5mL
Color Colorless to yellow
Density 1.15g/mL
Molecular Formula C5H9Cl2NSi
Synonym Dichloro(3-cyanopropyl)methylsilane
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Packaging Glass bottle
Chemical Name or Material Cyanopropylmethyldichlorosilane
Refractive Index 1.4570 to 1.4590 (20°C, 589nm)
Assay Percent Range 97%
CAS 1190-16-5
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.