Please login to your online account to display your discounted pricing

Cyanopindolol hemifumarate, Tocris

5-HT1A/1B antagonist; Also β-adrenergic antagonist

$188.60 - $794.96

Chemical Identifiers

Chemical Name or Material Cyanopindolol hemifumarate
Assay Percent Range >98%
Molecular Formula C16H21N3O2·1/2C4H4O4
Formula Weight 345.4
Synonym 4-[3-[tert-Butylamino]-2-hydroxypropoxy]-1H-indole-2-carbonitrile hemifumarate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

099310

 
r & d systems
099310
10mg Each for $188.60

099350

 
r & d systems
099350
50mg Each for $794.96
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 345.4
CAS 874882-72-1
Molecular Formula C16H21N3O2·1/2C4H4O4
Synonym 4-[3-[tert-Butylamino]-2-hydroxypropoxy]-1H-indole-2-carbonitrile hemifumarate
Assay Percent Range >98%
Chemical Name or Material Cyanopindolol hemifumarate
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature