Please login to your online account to display your discounted pricing

Alfa Aesar™ (±)-Cyanopindolol hemifumarate salt, 99%


Quantity 10mg

Manufacturer: alfa aesar™  J67445MA

 View more versions of this product

Catalog No. AAJ67445MA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Melting Point 250° to 252°C
Formula Weight 345.39
Chemical Name or Material (±)-Cyanopindolol hemifumarate salt
CAS 69906-85-0
MDL Number MFCD28100500
Synonym (±)-4-(3-tert-Butylamino-2-hydroxypropoxy)indole-2-carbonitrile hemifumarate
Physical Form Solid
Color White
Quantity 10mg
Assay 99%

Specifications

Chemical Name or Material (±)-Cyanopindolol hemifumarate salt
CAS 69906-85-0
Formula Weight 345.39
MDL Number MFCD28100500
Synonym (±)-4-(3-tert-Butylamino-2-hydroxypropoxy)indole-2-carbonitrile hemifumarate