Please login to your online account to display your discounted pricing

Alfa Aesar™ (±)-Cyanopindolol hemifumarate salt, 99%

Chemical Identifiers

Chemical Name or Material (±)-Cyanopindolol hemifumarate salt
CAS 69906-85-0
Formula Weight 345.39
MDL Number MFCD28100500
Synonym (±)-4-(3-tert-Butylamino-2-hydroxypropoxy)indole-2-carbonitrile hemifumarate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67445MA

 
alfa aesar™
J67445MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 250° to 252°C
Formula Weight 345.39
Chemical Name or Material (±)-Cyanopindolol hemifumarate salt
CAS 69906-85-0
MDL Number MFCD28100500
Synonym (±)-4-(3-tert-Butylamino-2-hydroxypropoxy)indole-2-carbonitrile hemifumarate
Physical Form Solid
Color White
Quantity 10mg
Assay 99%